Learn More
Newcomer Supply Michel Transport Medium Vials 20 ml vial, 10 ml fill, 25/case
SDP
Catalog No. NC2101910
Encompass
Use: Preserves morphology & antigenicity during transport of tissues. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.
This item is not returnable. View return policy
Catalog No. NC2101910 Supplier Newcomer Supply Supplier No. 12423E
Use: Preserves morphology & antigenicity during transport of tissues. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.