Learn More
Newcomer Supply Michel Transport Medium Vials 30 ml vial, 15 ml fill, 50/case
SDP
Catalog No. NC2101909
Encompass
Use: Preserves morphology & antigenicity during transport of tissues. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.
This item is not returnable. View return policy
Catalog No. NC2101909 Supplier Newcomer Supply Supplier No. 12423C
Use: Preserves morphology & antigenicity during transport of tissues. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.