Learn More
Stoelting™ Michele Trephine
SDP
Catalog No. 10001819 Shop All Stoelting Co Products
Change view
Click to view available options
Size:
3/16 in. OD
Catalog No. Size
10-001-819 3/16 in. OD
1 options
Catalog No. 10-001-819 Supplier Stoelting™ Supplier No. 5217004
Add to Cart
Add to Cart
Bone cutting tools allow for sharp cuts through soft bone and cartilage.
Specifications
For Use With (Application) Neuroscience
Length (English) 6.25 in.
Size 3/16 in. OD