Learn More
Cayman Chemical Micheliolide, Purity: ≥98%, CAS: 68370-47-8, 10 mg
SDP
Catalog No. 502934974
Encompass_Preferred
A sesquiterpene lactone with diverse biological activities; an irreversible activator of PKM2 (EC50 = 6 nM); decreases viability of HL-60 cells at 5-20 µM; reduces M. tuberculosis-induced secretion of IL-1β and TNF-α, expression of COX-2, and production of NO in RAW 264.7 cells; decreases the severity of CIA in a mouse model of rheumatoid arthritis at 30 mg/kg
Catalog No. 50-293-4974 Supplier Cayman Chemical Supplier No. 2540210 MG
A sesquiterpene lactone with diverse biological activities; an irreversible activator of PKM2 (EC50 = 6 nM); decreases viability of HL-60 cells at 5-20 µM; reduces M. tuberculosis-induced secretion of IL-1β and TNF-α, expression of COX-2, and production of NO in RAW 264.7 cells; decreases the severity of CIA in a mouse model of rheumatoid arthritis at 30 mg/kg