Learn More
Selleck Chemical LLC Micheliolide S9309-1MG
SDP
Catalog No. 5000024546
Encompass_Preferred
Micheliolide a sesquiterpene lactone isolated from Michelia compressa and Michelia champaca inhibits various inflammatory response and may serve as a neuroprotective agent in neuroinflammation-related neurodegenerative disorders
Catalog No. 50-000-24546 Supplier Selleck Chemical LLC Supplier No. S93091MG
Micheliolide a sesquiterpene lactone isolated from Michelia compressa and Michelia champaca inhibits various inflammatory response and may serve as a neuroprotective agent in neuroinflammation-related neurodegenerative disorders