Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Michels Solution
SDP

Catalog No. 22021401
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-021-401 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-401 Supplier StatLab Supplier No. FXMTMLT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

StatLab Michels Solution - used to preserve and transport specimens for examination in immunofluorescence studies

  • While not a fixative, specimens can be held at room temperature for five days in transport medium before processing
  • This simple salt solution maintains pH but does not kill most pathogens
  • Specimens received in transport medium should be washed in three changes of Michels Wash Solution (10 minutes for each wash)
  • Not suitable for FISH studies

Specifications

For Use With (Equipment) All Tissue Processors
Grade Histological
Quantity 1 L
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.