Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Michels Solution, StatLab™
GSA_VASDP

Catalog No. STLMS0507
Change view
Click to view available options
Quantity:
100 x 7 mL
96 x 20 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STLMS0507 96 x 20 mL
STLMS0735 100 x 7 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, if in contact with skin, or if inhaled.
Catalog No. STLMS0507 Supplier SLMP Llc Dba Statlab Medical Supplier No. MS0507
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Harmful if swallowed, if in contact with skin, or if inhaled.

Michels Solution is used to preserve and transport specimens for examination in immunofluorescence studies.

  • While not a fixative, specimens can be held at room temperature for five days in transport medium before processing
  • Simple salt solution maintains pH, but does not kill most pathogens
  • Specimens received in transport medium should be washed in three changes of Michels Wash Solution (10 min for each wash)
  • Not suitable for FISH studies

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Packaging Vial
Quantity 96 x 20 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.