Learn More
StatLab Michels Solution Transport Medium
SDP
Catalog No. 21033372
Change view
Click to view available options
Form:
Solution
Catalog No. Form
21-033-372 Solution
1 options
Catalog No. 21-033-372 Supplier StatLab Supplier No. FXMTMPT
Add to Cart
Add to Cart
Used to preserve and transport specimens for examination in immunofluorescence studies
  • While not a fixative, specimens can be held at room temperature for five days in transport medium before processing
  • This simple salt solution maintains pH but does not kill most pathogens
  • Specimens received in transport medium should be washed in three changes of Michels Wash Solution (Cat MS-32), 10 minutes for each wash
  • Not suitable for FISH studies
Specifications
Form Solution
Product Type Specimen Transport Medium
Serum Level Serum-free
Sterility Non-sterile
Sterilization Method Non-sterile