Learn More
Michels Wash Solution, StatLab™
GSA_VASDP
Catalog No. STLMS32
Change view
Click to view available options
Quantity:
32 oz.
Catalog No. Quantity
STLMS32 32 oz.
1 options
This item is not returnable. View return policy
Harmful if swallowed, if in contact with skin, or if inhaled.
Catalog No. STLMS32 Supplier SLMP Llc Dba Statlab Medical Supplier No. MS32
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, if in contact with skin, or if inhaled.
Used to wash specimens received in Michels transport medium.
  • Specimens should be washed in three changes of Michels washing solution
  • 10 minutes for each wash
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Packaging Bottle
Quantity 32 oz.
Concentration or Composition (by Analyte or Components) Water, Selected Buffers