Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Michler's Ketone, 98%

Catalog No. p-7081760
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1188922 100 g
AAA1188936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1188922 Supplier Thermo Scientific Chemicals Supplier No. A1188922
Only null left
Add to Cart

CAS: 90-94-8 | C17H20N2O | 268.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-94-8
Molecular Formula C17H20N2O
Molecular Weight (g/mol) 268.36
MDL Number MFCD00008312
InChI Key VVBLNCFGVYUYGU-UHFFFAOYSA-N
Synonym michler's ketone, 4,4'-bis dimethylamino benzophenone, michler ketone, bis 4-dimethylamino phenyl methanone, p,p'-michler's ketone, michlers ketone, methanone, bis 4-dimethylamino phenyl, tetramethyldiaminobenzophenone, p,p'-bis dimethylamino benzophenone
PubChem CID 7031
ChEBI CHEBI:82347
IUPAC Name bis[4-(dimethylamino)phenyl]methanone
SMILES CN(C)C1=CC=C(C=C1)C(=O)C1=CC=C(C=C1)N(C)C

Specifications

Melting Point 174°C to 176°C
Boiling Point >360°C
Flash Point 220°C (428°F)
Quantity 100 g
Beilstein 790733
Merck Index 14,6177
Formula Weight 268.36
Percent Purity 98%
Chemical Name or Material Michler's Ketone
Hazard Category H318-H341-H350
Hazard Statement GHS H Statement
H350-H341-H318
May cause cancer.
Suspected of causing genetic defects.
Causes serious eye damage.
Precautionary Statement P201-P202-P281-P305+P351+P338-P308+P313-P310-P501c
EINECSNumber 202-027-5
RTECSNumber DJ0250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.