Learn More
TARGETMOL CHEMICALS INC MICLXIN 5MG
SDP
Catalog No. 502579209
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Miclxin (DS37262926) is a novel MIC60 inhibitor that induces apoptosis through mitochondrial stress in mutant tumor cells via (beta)-catenin.Miclxin is a potent inhibitor of mutant (beta)-catenin which is involved in the Wnt signaling pathway.Miclxin induces (beta)-catenin-dependent apoptosis leading to mitochondrial damage and loss of mitochondrial membrane. Miclxin has anti-tumor activity and kills tumors by targeting MIC60. purity: 99%
Catalog No. 50-257-9209 Supplier TARGETMOL CHEMICALS INC Supplier No. T734155MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Miclxin (DS37262926) is a novel MIC60 inhibitor that induces apoptosis through mitochondrial stress in mutant tumor cells via (beta)-catenin.Miclxin is a potent inhibitor of mutant (beta)-catenin which is involved in the Wnt signaling pathway.Miclxin induces (beta)-catenin-dependent apoptosis leading to mitochondrial damage and loss of mitochondrial membrane. Miclxin has anti-tumor activity and kills tumors by targeting MIC60. purity: 99%