Learn More
Alkali Scientific Miconazole, 25 G
SDP
Catalog No. 5000659642
Encompass_Preferred
Miconazole, 25 g, is a potent antifungal agent widely used in laboratory settings for antifungal susceptibility testing and research. Miconazole functions by inhibiting the synthesis of ergosterol in fungal cell membranes, leading to cell membrane disruption and fungal cell death. It is effective against a broad spectrum of fungi, including Candida species, dermatophytes, and other pathogenic fungi. The 25 g quantity is often used for bulk studies, including determining minimum inhibitory concentrations (MICs) for miconazole in combination with other antifungal agents. Researchers also utilize miconazole in exploring new antifungal treatments and in studying resistance mechanisms in fungal pathogens.
Catalog No. 50-006-59642 Supplier Alkali Scientific Supplier No. ICO002CP
Miconazole, 25 g, is a potent antifungal agent widely used in laboratory settings for antifungal susceptibility testing and research. Miconazole functions by inhibiting the synthesis of ergosterol in fungal cell membranes, leading to cell membrane disruption and fungal cell death. It is effective against a broad spectrum of fungi, including Candida species, dermatophytes, and other pathogenic fungi. The 25 g quantity is often used for bulk studies, including determining minimum inhibitory concentrations (MICs) for miconazole in combination with other antifungal agents. Researchers also utilize miconazole in exploring new antifungal treatments and in studying resistance mechanisms in fungal pathogens.