Learn More
Alkali Scientific Miconazole, 5 G
SDP
Catalog No. 5000659682
Encompass_Preferred
Miconazole, 5 g, is an antifungal reagent commonly used in laboratory research to investigate fungal infections and study antifungal resistance. This imidazole-based antifungal agent works by inhibiting the synthesis of ergosterol, a vital component of the fungal cell membrane, resulting in membrane disruption and cell death. Miconazole is particularly effective against Candida species, dermatophytes, and other pathogenic fungi. The 5 g size is ideal for testing the compound’s effectiveness in laboratory assays, including susceptibility testing, MIC determinations, and studies examining fungal resistance. Researchers also use miconazole in the development of new antifungal therapies and understanding resistance mechanisms in clinical isolates.
Catalog No. 50-006-59682 Supplier Alkali Scientific Supplier No. ICO003CP
Miconazole, 5 g, is an antifungal reagent commonly used in laboratory research to investigate fungal infections and study antifungal resistance. This imidazole-based antifungal agent works by inhibiting the synthesis of ergosterol, a vital component of the fungal cell membrane, resulting in membrane disruption and cell death. Miconazole is particularly effective against Candida species, dermatophytes, and other pathogenic fungi. The 5 g size is ideal for testing the compound’s effectiveness in laboratory assays, including susceptibility testing, MIC determinations, and studies examining fungal resistance. Researchers also use miconazole in the development of new antifungal therapies and understanding resistance mechanisms in clinical isolates.