Learn More
Medchemexpress LLC Miconazole (nitrate) | 22832-87-7 | >99.4% | 25 MG
SDP
Catalog No. 5000451014
Encompass_Preferred
Miconazole nitrate is an imidazole antifungal agent that can cross the blood-brain barrier and also possesses antibacterial effects. It functions by inhibiting the synthesis of ergosterol, a crucial component of fungal cell membranes, making it effective against certain species of Leishmania protozoa.
  • Imidazole antifungal agent
  • Crosses the blood-brain barrier
  • Possesses antibacterial effects
  • Inhibits ergosterol synthesis
  • Effective against Leishmania protozoa
  • Used in Ektachrome film developing
Catalog No. 50-004-51014 Supplier Medchemexpress LLC Supplier No. 25MGHYB0454AR
Miconazole nitrate is an imidazole antifungal agent that can cross the blood-brain barrier and also possesses antibacterial effects. It functions by inhibiting the synthesis of ergosterol, a crucial component of fungal cell membranes, making it effective against certain species of Leishmania protozoa.
  • Imidazole antifungal agent
  • Crosses the blood-brain barrier
  • Possesses antibacterial effects
  • Inhibits ergosterol synthesis
  • Effective against Leishmania protozoa
  • Used in Ektachrome film developing