Learn More
Medchemexpress LLC Miconazole nitrate | 22832-87-7 | 99.98% | 250 MG
SDP
Catalog No. 5000407050
Encompass_Preferred
Miconazole nitrate is an imidazole antifungal agent that can pass through the blood-brain barrier and also exhibits antibacterial effects. Developed by Janssen Pharmaceutica, it is commonly applied topically to treat fungal infections by inhibiting the synthesis of ergosterol, a crucial component of fungal cell membranes. It is also effective against certain species of Leishmania protozoa and possesses antiparasitic properties.
  • Imidazole antifungal agent
  • Can pass through the blood-brain barrier
  • Exhibits antibacterial effects
  • Inhibits the synthesis of ergosterol in fungal cell membranes
  • Effective against certain species of Leishmania protozoa
  • Applied topically to cure fungal infections
  • Used in Ektachrome film developing
  • Included as a final rinse additive in rapid access color negative processes
Catalog No. 50-004-07050 Supplier Medchemexpress LLC Supplier No. 250MGHYB0454AR
Add to Cart
Add to Cart
Miconazole nitrate is an imidazole antifungal agent that can pass through the blood-brain barrier and also exhibits antibacterial effects. Developed by Janssen Pharmaceutica, it is commonly applied topically to treat fungal infections by inhibiting the synthesis of ergosterol, a crucial component of fungal cell membranes. It is also effective against certain species of Leishmania protozoa and possesses antiparasitic properties.
  • Imidazole antifungal agent
  • Can pass through the blood-brain barrier
  • Exhibits antibacterial effects
  • Inhibits the synthesis of ergosterol in fungal cell membranes
  • Effective against certain species of Leishmania protozoa
  • Applied topically to cure fungal infections
  • Used in Ektachrome film developing
  • Included as a final rinse additive in rapid access color negative processes