Learn More
Alkali Scientific Miconazole Nitrate, 25 G
Supplier: Alkali Scientific MIC001CP
Miconazole Nitrate, a potent antifungal agent, is provided in a 25g powder form for research and clinical applications. It is widely used in the treatment of fungal infections, particularly those caused by dermatophytes and yeasts such as Candida species. Miconazole works by inhibiting ergosterol synthesis, an essential component of fungal cell membranes, leading to cell death. This compound is often used in laboratory experiments focusing on antifungal drug development, resistance studies, and the mechanism of fungal infection. The 25g amount allows for the preparation of large batches for in vitro testing, particularly in pharmacological and microbiological research.