Learn More
Selleck Chemical LLC Miconazole Nitrate S1956-50mg
SDP
Catalog No. 503109027
Encompass_Preferred
Miconazole (NSC 169434) Nitrate is an imidazole antifungal agent by inhibiting ergosterol biosynthesis and inducing ROS used to treat vaginal yeast infections
Catalog No. 50-310-9027 Supplier Selleck Chemical LLC Supplier No. S195650MG
Miconazole (NSC 169434) Nitrate is an imidazole antifungal agent by inhibiting ergosterol biosynthesis and inducing ROS used to treat vaginal yeast infections