Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-Miconazole, Nitrate, MP Biomedicals™

Catalog No. ICN15169380 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15169380 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15169380 Supplier MP Biomedicals, Inc Supplier No. 0215169380
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 75319-48-1
Color White
Molecular Formula C18H15Cl4N3O4
Quantity 100 g
Synonym miconazole nitrate, albistat, andergin, aflorix, conofite, florid, micatin, gyno-monistat, epi-monistat, gyno-daktar
InChI Key MCCACAIVAXEFAL-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1Cl)Cl)COC(CN2C=CN=C2)C3=C(C=C(C=C3)Cl)Cl.[N+](=O)(O)[O-]
IUPAC Name 1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]imidazole;nitric acid
Molecular Weight (g/mol) 479.135
PubChem CID 68553
Formula Weight 479.1
Physical Form Powder
Chemical Name or Material (±)-Miconazole
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.