Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-Miconazole nitrate salt, Thermo Scientific Chemicals

Catalog No. AC291830010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC291830010 1 g
AC291830050 5 g
AC291830250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC291830010 Supplier Thermo Scientific Chemicals Supplier No. 291830010
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22832-87-7
Molecular Formula C18H15Cl4N3O4
Molecular Weight (g/mol) 479.14
MDL Number MFCD00058161
InChI Key MCCACAIVAXEFAL-UHFFFAOYNA-N
PubChem CID 68553
IUPAC Name 1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]imidazole;nitric acid
SMILES O[N+]([O-])=O.ClC1=CC=C(COC(CN2C=CN=C2)C2=CC=C(Cl)C=C2Cl)C(Cl)=C1

Specifications

Type (±)-Miconazole Nitrate Salt
Infrared Spectrum Authentic
Loss on Drying 0.5% max. (105°C, 2 h)
Quantity 1 g
Solubility Information Solubility in water: slightly soluble.
Formula Weight 479.14
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material (±)-Miconazole nitrate salt
Hazard Category Warning
Hazard Statement GHS H Statement Harmful if swallowed. May cause an allergic skin reaction.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. If skin irritation or rash occurs: Get medical advice/attention. Wash contaminated clothing before reuse.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.