Learn More
Research Products International Corp Micrewtube Screw Cap Tube with Loop, Non-Graduated, 2.0ml Conical Bottom, 500 per Case
SDP
Catalog No. 501367506
Encompass
Micrewtube Tubes features O-ring seal screw cap and attachment loop. Loops remain attached to tubes and prevent cross contamination. Tubes withstand high speed centrifuging, autoclaving, boiling and freezer storage. Screw cap with "O" ring provides superior sealing during sample processing. Uniform well thickness withstands 17,000xg forces. Graduated tubes have printed marking area. Polypropylene tubes are available with screwed-on cap: pre-attached loop; natural color; and sterile.
Catalog No. 50-136-7506 Supplier Research Products International Corp Supplier No. 144533
Micrewtube Tubes features O-ring seal screw cap and attachment loop. Loops remain attached to tubes and prevent cross contamination. Tubes withstand high speed centrifuging, autoclaving, boiling and freezer storage. Screw cap with "O" ring provides superior sealing during sample processing. Uniform well thickness withstands 17,000xg forces. Graduated tubes have printed marking area. Polypropylene tubes are available with screwed-on cap: pre-attached loop; natural color; and sterile.