Learn More
Andwin Scientific MICRFNL SP .2UM WHT GRID 50/PK
SDP
Catalog No. NC9020852
Encompass_Preferred
MicroFunnel Disposable Filter Funnel, Sterile, w/0.2um supor polyethersulfone Membrane. White gridded. Filtration area 12.25cm2. 635mmHg vacuum. 5.74 dia.x7.32 cm H, w/o lid, 100mL
Catalog No. NC9020852 Supplier Andwin Scientific Supplier No. 4803
MicroFunnel Disposable Filter Funnel, Sterile, w/0.2um supor polyethersulfone Membrane. White gridded. Filtration area 12.25cm2. 635mmHg vacuum. 5.74 dia.x7.32 cm H, w/o lid, 100mL