Learn More
DUBOIS CHEMICALS MICRO 90 5 GALLON
Catalog No. NC9405454
Encompass
Micro-90� is a mild, yet powerful multipurpose, alkaline cleaning concentrate that has long been used for laboratory cleaning, industrial applications, and critical cleaning processes. Micro-90 is a unique formulation of chelants, ionic, and non-ionic ingredients that produce a variety of cleaning actions that include lifting, dispersing, emulsifying, sequestering, suspending, and decomposing. Target soils for Micro-90 include oil, grease, wax, tar, flux, particulates, hard water stains, and biological debris. Micro-90 is highly effective at defouling filter membranes and can be validated in critical cleaning applications. Micro-90 Wash Methods * CIP * Immersion * Manual * Mild-agitation * Ultrasonic
Catalog No. NC9405454 Supplier DUBOIS CHEMICALS Supplier No. M9033
Micro-90� is a mild, yet powerful multipurpose, alkaline cleaning concentrate that has long been used for laboratory cleaning, industrial applications, and critical cleaning processes. Micro-90 is a unique formulation of chelants, ionic, and non-ionic ingredients that produce a variety of cleaning actions that include lifting, dispersing, emulsifying, sequestering, suspending, and decomposing. Target soils for Micro-90 include oil, grease, wax, tar, flux, particulates, hard water stains, and biological debris. Micro-90 is highly effective at defouling filter membranes and can be validated in critical cleaning applications. Micro-90 Wash Methods * CIP * Immersion * Manual * Mild-agitation * Ultrasonic