Learn More
Andwin Scientific MICRO 90 CLEAN SOLN-CASE/4 4L
SDP
Catalog No. NC0167017
Encompass_Preferred
The Micro-90 Concentrated Alkaline Cleaning Solution is a powerful cleaning concentrate that can efficiently remove oil, grease, wax, tar, flux, particulates, hard water stains, and biological debris. This solution is composed of chelants, ionic, and non-ionic ingredients that generate many cleaning actions such as lifting, dispersing, emulsifying, sequestering, suspending, and decomposing. This product is ideal for laboratory cleaning, industrial applications, and critical cleaning processes. It is packaged in a 4 liter plastic bottle and is sold in a case of 4.
Catalog No. NC0167017 Supplier Andwin Scientific Supplier No. 89177070
The Micro-90 Concentrated Alkaline Cleaning Solution is a powerful cleaning concentrate that can efficiently remove oil, grease, wax, tar, flux, particulates, hard water stains, and biological debris. This solution is composed of chelants, ionic, and non-ionic ingredients that generate many cleaning actions such as lifting, dispersing, emulsifying, sequestering, suspending, and decomposing. This product is ideal for laboratory cleaning, industrial applications, and critical cleaning processes. It is packaged in a 4 liter plastic bottle and is sold in a case of 4.