Learn More
Andwin Scientific MICRO-90 CLEANING SOL'N 12X1QT
SDP
Catalog No. NC9549180
Encompass_Preferred
The Micro-90� Concentrated Alkaline Cleaning Solution is a powerful cleaning concentrate that can efficiently remove oil, grease, wax, tar, flux, particulates, hard water stains, and biological debris. This solution is composed of chelants, ionic, and non-ionic ingredients that generate many cleaning actions such as lifting, dispersing, emulsifying, sequestering, suspending, and decomposing. This product is ideal for laboratory cleaning, industrial applications, and critical cleaning processes. It is packaged and sold in a 1 quart plastic bottle.
Catalog No. NC9549180 Supplier Andwin Scientific Supplier No. 9031
The Micro-90� Concentrated Alkaline Cleaning Solution is a powerful cleaning concentrate that can efficiently remove oil, grease, wax, tar, flux, particulates, hard water stains, and biological debris. This solution is composed of chelants, ionic, and non-ionic ingredients that generate many cleaning actions such as lifting, dispersing, emulsifying, sequestering, suspending, and decomposing. This product is ideal for laboratory cleaning, industrial applications, and critical cleaning processes. It is packaged and sold in a 1 quart plastic bottle.