Learn More
Andwin Scientific MICRO-90 CLEANING SOL'N 1L
SDP
Catalog No. NC0892451
Encompass_Preferred
The Micro-90� Concentrated Alkaline Cleaning Solution is a powerful cleaning concentrate that can efficiently remove oil, grease, wax, tar, flux, particulates, hard water stains, and biological debris. This solution is composed of chelants, ionic, and non-ionic ingredients that generate many cleaning actions such as lifting, dispersing, emulsifying, sequestering, suspending, and decomposing. This product is ideal for laboratory cleaning, industrial applications, and critical cleaning processes. It is packaged and sold in a 1-liter plastic bottle.
Catalog No. NC0892451 Supplier Andwin Scientific Supplier No. 89210138EA
The Micro-90� Concentrated Alkaline Cleaning Solution is a powerful cleaning concentrate that can efficiently remove oil, grease, wax, tar, flux, particulates, hard water stains, and biological debris. This solution is composed of chelants, ionic, and non-ionic ingredients that generate many cleaning actions such as lifting, dispersing, emulsifying, sequestering, suspending, and decomposing. This product is ideal for laboratory cleaning, industrial applications, and critical cleaning processes. It is packaged and sold in a 1-liter plastic bottle.