Learn More
Andwin Scientific MICRO-90 CLEANING SOLUTION 1GL
SDP
Catalog No. NC0518433
Encompass_Preferred
The Micro-90 Concentrated Alkaline Cleaning Solution is a powerful cleaning concentrate that can efficiently remove oil, grease, wax, tar, flux, particulates, hard water stains, and biological debris. This solution is composed of chelants, ionic, and non-ionic ingredients that generate many cleaning actions such as lifting, dispersing, emulsifying, sequestering, suspending, and decomposing. This product is ideal for laboratory cleaning, industrial applications, and critical cleaning processes. It is packaged and sold in a 1 gallon plastic bottle.
Catalog No. NC0518433 Supplier Andwin Scientific Supplier No. 5409B
The Micro-90 Concentrated Alkaline Cleaning Solution is a powerful cleaning concentrate that can efficiently remove oil, grease, wax, tar, flux, particulates, hard water stains, and biological debris. This solution is composed of chelants, ionic, and non-ionic ingredients that generate many cleaning actions such as lifting, dispersing, emulsifying, sequestering, suspending, and decomposing. This product is ideal for laboratory cleaning, industrial applications, and critical cleaning processes. It is packaged and sold in a 1 gallon plastic bottle.