Learn More
Beckman Coulter MICRO-90 Neutral Detergent
Catalog No. 15655223
Change view
Click to view available options
Quantity:
0.946 L
Catalog No. Quantity
15-655-223 0.946 L
1 options
Catalog No. 15-655-223 Supplier Beckman Coulter Supplier No. 8304096/DEL
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
MICRO-90 Neutral Detergent should be used primarily to clean inside surfaces of liquid modules. MICRO-90 can also be used as dispersant.
  • Recommended for cleaning certain parts of the LS 13 320 Series and Multisizer Instruments
  • Multi-purpose liquid for cleaning glassware, lab equipment, and other surfaces
TRUSTED_SUSTAINABILITY
Specifications
Product Type Detergent
Capacity (Metric) 0.946 L
Quantity 0.946 L
For Use With (Application) Cleaning Inside Surfaces of Liquid Modules