Learn More
Cole-Parmer International Products Corp M-0801-12 Micro A07 Citric Acid Cleaner, 12/cs
Catalog No. NC1622079 Shop All Cole-Parmer Products
Encompass_Preferred
Micro A07 is a free-rinsing cleaner designed for use in a wide range of applications including ultrasonic baths, immersion tanks, mild-agitation systems, CIP systems and more. It is ideal for cleaning metals, stainless steel, ceramics, glass, plastics, and filter membranes. Micro A07 solution removes a variety of soils as well including scale, oxides, milkstone, mineral deposits, and inorganic materials. It is free of CFCs, ODCs, phosphates, silicates, borates, halogenated compounds, and phenols.
Catalog No. NC1622079 Supplier Cole-Parmer Supplier No. 1810401
Micro A07 is a free-rinsing cleaner designed for use in a wide range of applications including ultrasonic baths, immersion tanks, mild-agitation systems, CIP systems and more. It is ideal for cleaning metals, stainless steel, ceramics, glass, plastics, and filter membranes. Micro A07 solution removes a variety of soils as well including scale, oxides, milkstone, mineral deposits, and inorganic materials. It is free of CFCs, ODCs, phosphates, silicates, borates, halogenated compounds, and phenols.