Learn More
Fine Science Tools Micro-Adson Forceps, Fenestrated Handle, 12 cm
SDP
Catalog No. NC0584641
Encompass
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 0.8 x 0.9mm/ Alloy / Material: Stainless Steel/ Length: 12cm
Catalog No. NC0584641 Supplier Fine Science Tools Supplier No. 1102012
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 0.8 x 0.9mm/ Alloy / Material: Stainless Steel/ Length: 12cm