Learn More
Stoelting™ Micro Adson Forceps
SDP
Catalog No. 10001221 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Serrated
Catalog No. Model
10-001-221 Serrated
1 options
Catalog No. 10-001-221 Supplier Stoelting™ Supplier No. 5210439
Add to Cart
Add to Cart
Stoelting™ Micro Adson Tissue Forceps are micro tweezers that are useful in a variety of different applications including micro dissection of tissue and vessels, and processing tissue in electron microscopy procedures.
Micro tweezers come in a wide variety of tip sizes, shapes, and materials. To choose the best tweezers, keep in mind the size of the animal and/or tissue. The type of cleaning and sterilization required may also affect the chosen material. If autoclaving is required or users are working under a microscope on a mouse, the thinnest-tipped forceps made of stainless steel, titanium, Dumostar, INOX, and Dumoxel are good choices.
Carbon steel instruments may not be the best choice for use where there is constant exposure to water. If not handled properly, carbon steel is likely to rust if exposed to water for a length of time. Though not recommended, flash flame decontamination could be used on carbon steel tweezers because this material is less likely to be damaged by high heat. The extreme heat from flash flame decontamination changes the molecular structure of most material adversely. High heat softens metal and shortens the useful life of instruments. Consider another method of decontamination where lower heat levels can be applied to keep your instruments useful for many more years.
Specifications
For Use With (Application) Neuroscience
Length (English) 4.75 in.
Width (Metric) 1 mm tip
Model Serrated