Learn More
Roboz Surgical MICRO ANEUR CLIP
SDP
Catalog No. NC9202107
Encompass
Micro Clip; Straight; 70 Grams Pressure; 0.75mm Clip Width; 5mm Jaw Length. The jaws have fine atraumatic serrations. Please use RS-5410 or RS-5440 clip applying forceps with these clips. Compressing the clips with fingers is very difficult and
Catalog No. NC9202107 Supplier Roboz Surgical Supplier No. RS5420
Micro Clip; Straight; 70 Grams Pressure; 0.75mm Clip Width; 5mm Jaw Length. The jaws have fine atraumatic serrations. Please use RS-5410 or RS-5440 clip applying forceps with these clips. Compressing the clips with fingers is very difficult and