Learn More
World Precision Instrument Micro-Applicator Stick, 2.0mm Tip, Pack of 100
SDP
Catalog No. 502538404
Encompass
These flexible applicator brushes have unique, non-absorbent, synthetic fibers. Solution is held in suspension until applied - eliminating dripping, spilling and waste. Great for solvents, adhesives and enamels. Applicator brush ends can be bent to any angle for precise application (choose brush diameter of 1.50mm, 2.00mm or 2.50mm). Sold in package of 100.
Catalog No. 50-253-8404 Supplier World Precision Instrument Supplier No. W16104
These flexible applicator brushes have unique, non-absorbent, synthetic fibers. Solution is held in suspension until applied - eliminating dripping, spilling and waste. Great for solvents, adhesives and enamels. Applicator brush ends can be bent to any angle for precise application (choose brush diameter of 1.50mm, 2.00mm or 2.50mm). Sold in package of 100.