Learn More
World Precision Instrument Micro Applicator Stick, 2.5mm Tip, Pack of 100
SDP
Catalog No. 502538403
Encompass
Disposable micro applicators have non-linting, non-absorbent fibers arranged in a cone shape.  They hold solutions in suspension, eliminating dripping.  The flexible portion allows for precise application in hard to reach areas.  Brush fiber section is approximately 2.5mm x 1.5mm diameter.  Convenient plastic dispenser contains 100 applicators.
Catalog No. 50-253-8403 Supplier World Precision Instrument Supplier No. W16105
Disposable micro applicators have non-linting, non-absorbent fibers arranged in a cone shape.  They hold solutions in suspension, eliminating dripping.  The flexible portion allows for precise application in hard to reach areas.  Brush fiber section is approximately 2.5mm x 1.5mm diameter.  Convenient plastic dispenser contains 100 applicators.