Learn More
Perkin Elmer US LLC Micro ATR Objective for the Multiscope Systems
Catalog No. 502117254
Encompass_Preferred
This accessory offers transmission, reflectance, and visible viewing modes all in one device; no realignment or extra equipment required.
Catalog No. 50-211-7254 Supplier Perkin Elmer US LLC Supplier No. L1860298
Add to Cart
Add to Cart
Request a Quote
This accessory offers transmission, reflectance, and visible viewing modes all in one device; no realignment or extra equipment required.