Learn More
Perkin Elmer US LLC Micro ATR Objective Upgrade Kit
Catalog No. 502117262
Encompass_Preferred
This kit includes a Cassegrain Reflector, a mount and a Germanium (GE) micro ATR crystal with holder. A spare Germanium crystal and holder are also included.
Catalog No. 50-211-7262 Supplier Perkin Elmer US LLC Supplier No. L1860334
Add to Cart
Add to Cart
Request a Quote
This kit includes a Cassegrain Reflector, a mount and a Germanium (GE) micro ATR crystal with holder. A spare Germanium crystal and holder are also included.