Learn More
Cayman Chemical Micro BCA Protein Assay Kit, 480 Well
SDP
Catalog No. 502894037
Encompass_Preferred
Cayman’s Micro BCA Protein Assay Kit is a quantitative assay that can be used with various buffers, water, plasma, serum, urine, and cell lysates. The assay has a dynamic range of 4 to 40 µg/ml, a lower limit detection (LLOD) of approximately 2 µg/ml, and a lower limit of quantification (LLOQ) of 4 µg/ml.
Catalog No. 50-289-4037 Supplier Cayman Chemical Supplier No. 760200480 WELL
Cayman’s Micro BCA Protein Assay Kit is a quantitative assay that can be used with various buffers, water, plasma, serum, urine, and cell lysates. The assay has a dynamic range of 4 to 40 µg/ml, a lower limit detection (LLOD) of approximately 2 µg/ml, and a lower limit of quantification (LLOQ) of 4 µg/ml.