Learn More
Thomas Scientific Refill Glass Beads, 3mm, 1000g
SDP
Catalog No. 501458145
Encompass_Preferred
Benchmark's Micro Bead Sterilizer is ideal for the sterilization of small research tools (forceps, scissors, etc.). When the chamber is filled with the included glass beads, high temperatures (up to 300°C) can be used to eliminate the presence of bacteria, spores and other microorganisms. Chamber temperature can be viewed on the large LED control panel and conveniently adjusted with the control knob. The outer casing of the unit stays cool to the touch, even when the sterilizer is used all day. The supplied safety cover can be placed over the opening to conserve energy and reduce the heating/ramping rate. The Micro Bead Sterilizer does not use gas, flame or chemicals and is safe for use in hoods. With adjustable temperature, the application range of the sterilizer can also be exanded into incubations and the decontamination of plastic objects/tools. Digital control, up to 300°C Sterilizes forceps, needles & other tools (generally in 15 sec.) Eliminates the danger associ
Catalog No. 50-145-8145 Supplier Thomas Scientific Supplier No. 1177X44
Benchmark's Micro Bead Sterilizer is ideal for the sterilization of small research tools (forceps, scissors, etc.). When the chamber is filled with the included glass beads, high temperatures (up to 300°C) can be used to eliminate the presence of bacteria, spores and other microorganisms. Chamber temperature can be viewed on the large LED control panel and conveniently adjusted with the control knob. The outer casing of the unit stays cool to the touch, even when the sterilizer is used all day. The supplied safety cover can be placed over the opening to conserve energy and reduce the heating/ramping rate. The Micro Bead Sterilizer does not use gas, flame or chemicals and is safe for use in hoods. With adjustable temperature, the application range of the sterilizer can also be exanded into incubations and the decontamination of plastic objects/tools. Digital control, up to 300°C Sterilizes forceps, needles & other tools (generally in 15 sec.) Eliminates the danger associ