Learn More
Fine Science Tools Micro Bead Sterilizer with Glass Beads
SDP
Catalog No. NC3826084
Encompass
Mico Bead Research Sterilizer with Glass Beads; 110V (suitable for US electrical plug) Micro Bead Sterilizer is ideal for the sterilization of small research tools (forceps, scissors, etc.). When the chamber is filled with the included glass beads, high temperatures (up to 300° C) can be used to eliminate the presence of bacteria, spores and other micro-organisms. The typical exposure time is about 10 seconds for sterilization of commonly used items, such as forceps, scissors, tweezers, scalpels, inoculation loops, etc.This product is designed for research use only.Not intended for clinical use. Digital control, up to 300° CIncludes Glass beads (150 g)Temperature range: 100 to 300° C
Catalog No. NC3826084 Supplier Fine Science Tools Supplier No. 1809045
Mico Bead Research Sterilizer with Glass Beads; 110V (suitable for US electrical plug) Micro Bead Sterilizer is ideal for the sterilization of small research tools (forceps, scissors, etc.). When the chamber is filled with the included glass beads, high temperatures (up to 300° C) can be used to eliminate the presence of bacteria, spores and other micro-organisms. The typical exposure time is about 10 seconds for sterilization of commonly used items, such as forceps, scissors, tweezers, scalpels, inoculation loops, etc.This product is designed for research use only.Not intended for clinical use. Digital control, up to 300° CIncludes Glass beads (150 g)Temperature range: 100 to 300° C