Learn More
Bio Rad Laboratories MICRO BIO-SPIN 30 COLUMNS
Catalog No. 509011913
Encompass
30 Columns; Bio-Rad; Micro Bio-Spin; Includes 100 columns in Tris buffer, 200 collection tubes, RNase-free
Catalog No. 50-901-1913 Supplier Bio Rad Laboratories Supplier No. 7326251
30 Columns; Bio-Rad; Micro Bio-Spin; Includes 100 columns in Tris buffer, 200 collection tubes, RNase-free