Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stoelting™ Micro Blumenthal Rongeur
SDP

Catalog No. 10000835 Shop All Stoelting Co Products
Change view
Click to view available options
Width (Metric):
1.3 mm
N/A
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (Metric)
10-000-835 1.3 mm
10-001-973 N/A
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-000-835 Supplier Stoelting™ Supplier No. 52161
Only null left
Add to Cart
Add to Cart

Rongeurs feature a sharp-edged, scoop-shaped tip, used for gouging out bone or opening a window in bone, often in the skull. It is used in neurosurgery and orthopedic surgery to expose areas for operation or to aid in removal of the brain.

Specifications

Length (English) 4.5 in.
Width (Metric) 1.3 mm
Model Curved
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.