Learn More
Stoelting™ Micro Blumenthal Rongeur
SDP
Catalog No. 10001973 Shop All Stoelting Co Products
Change view
Click to view available options
Width (Metric):
1.3 mm
N/A
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (Metric)
10-001-973 N/A
10-000-835 1.3 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-001-973 Supplier Stoelting™ Supplier No. 52164P
Add to Cart
Add to Cart
Rongeurs feature a sharp-edged, scoop-shaped tip, used for gouging out bone or opening a window in bone, often in the skull. It is used in neurosurgery and orthopedic surgery to expose areas for operation or to aid in removal of the brain.
Specifications
Length (English) 4.5 in.
Width (Metric) N/A
Model Curved