Learn More
General Laboratory Products MICRO BUFFER TW/LEC 490ML 12CS
Catalog No. 50821507
Encompass_Preferred
Microbiology supplies, Media for microbiology, Culture media for microbiology, General Laboratory Products, Phosphate Buffered Water w/ 4.0% Tween20 and 0.5 % Lecithin, 490 ML, W/M Jar, 12/CS
Catalog No. 50-821-507 Supplier General Laboratory Products Supplier No. 500EZ1170490
Microbiology supplies, Media for microbiology, Culture media for microbiology, General Laboratory Products, Phosphate Buffered Water w/ 4.0% Tween20 and 0.5 % Lecithin, 490 ML, W/M Jar, 12/CS