Learn More
Premier Lab Supply Micro Carbon Residue Testing Vials – PN# 915-4530-120
SDP
Catalog No. NC1995643
Encompass
Micro Carbon Residue Testing Vials holding CC: 0.5 Dram/2.0mL (PN# 915-4530-120) – Shell type, plain top, short style – Dimensions: OD 12mm, Length 35mm – Suitable for MCT; Method 4530 – These vials are disposable, economically priced and are fabricated from N-51A borosilicate glass. The walls are a straight-sided design that makes them ideal for micro carbon residue testing. – 100 per package
Catalog No. NC1995643 Supplier Premier Lab Supply Supplier No. 9154530120
Micro Carbon Residue Testing Vials holding CC: 0.5 Dram/2.0mL (PN# 915-4530-120) – Shell type, plain top, short style – Dimensions: OD 12mm, Length 35mm – Suitable for MCT; Method 4530 – These vials are disposable, economically priced and are fabricated from N-51A borosilicate glass. The walls are a straight-sided design that makes them ideal for micro carbon residue testing. – 100 per package