Learn More
World Precision Instrument Micro Castroviejo Needle Holder, 9cm, Straight, Diamond Dust Tips
SDP
Catalog No. 502539487
Encompass
Micro Castroviejo Needle Holders are a surgical instrument used for microsurgery requiring precise control and manipulation of small needles, though they are most commonly used in ophthalmic surgery. These have a fine rounded tip. Our Castroviejo Needle Holders feature fine tips for grasping and manipulation of small, delicate needles. The jaws of these Micro Castroviejo Needle Holders are diamond dusted for a secure grasp of fine needles. These Catroviejo needle holders have a spring action for situations when you need constant pressure on the needle and precision control. These are useful when working under a microscope or in confined spaces. They are an excellent choice for left-handed operators, too, as they are designed for ambidextrous use. Be sure to clean, disinfect, and sterilize your surgical instruments promptly after each use to ensure a long life for your surgical tools.
Catalog No. 50-253-9487 Supplier World Precision Instrument Supplier No. 503376
Micro Castroviejo Needle Holders are a surgical instrument used for microsurgery requiring precise control and manipulation of small needles, though they are most commonly used in ophthalmic surgery. These have a fine rounded tip. Our Castroviejo Needle Holders feature fine tips for grasping and manipulation of small, delicate needles. The jaws of these Micro Castroviejo Needle Holders are diamond dusted for a secure grasp of fine needles. These Catroviejo needle holders have a spring action for situations when you need constant pressure on the needle and precision control. These are useful when working under a microscope or in confined spaces. They are an excellent choice for left-handed operators, too, as they are designed for ambidextrous use. Be sure to clean, disinfect, and sterilize your surgical instruments promptly after each use to ensure a long life for your surgical tools.