Learn More
Azzota Corp Micro cell frosted lid 10mm 1.
Catalog No. 5000886973
Encompass_Preferred
Micro Cell with Frosted Walls and Lid Outside Dimensions 45x12 5x12 5mm Path Length 10mm Inside Width 4mm 1 4ml Volume
Catalog No. 50-008-86973 Supplier Azzota Corp Supplier No. G264
Micro Cell with Frosted Walls and Lid Outside Dimensions 45x12 5x12 5mm Path Length 10mm Inside Width 4mm 1 4ml Volume