Learn More
Perkin Elmer US LLC Micro Cell Holder for LAMBDA 365
Catalog No. 502105531
Encompass_Preferred
PerkinElmer provides optional cell holders and changers that make it possible for you to measure samples from thin films to highly-diffuse powders at fixed or variable angles. With these specialty cell holders, you can extend the types of samples you analyze as well as the types of analysis you conduct.
Catalog No. 50-210-5531 Supplier Perkin Elmer US LLC Supplier No. N4101028
PerkinElmer provides optional cell holders and changers that make it possible for you to measure samples from thin films to highly-diffuse powders at fixed or variable angles. With these specialty cell holders, you can extend the types of samples you analyze as well as the types of analysis you conduct.