Learn More
Starna Cells Inc MICRO CELLS .700ML
SDP
Catalog No. NC9676929
Encompass_Preferred
Spectrosil Far UV Quartz windows Useable range: 170 to 2700nm Pathlength: 10mm Exterior Width: 12.5mm Exterior Length: 12.5mm Exterior Height: 45mm Interior Width: 2mm Interior Length: 10mm Nominal Volume: 0.7mL Number polished windows:2
Catalog No. NC9676929 Supplier Starna Cells Inc Supplier No. 18Q10
Spectrosil Far UV Quartz windows Useable range: 170 to 2700nm Pathlength: 10mm Exterior Width: 12.5mm Exterior Length: 12.5mm Exterior Height: 45mm Interior Width: 2mm Interior Length: 10mm Nominal Volume: 0.7mL Number polished windows:2