Learn More
CELLTREAT Scientific Products 2.0mL Micro Centrifuge Tube, Low Retention, Non-sterile
SDP
Catalog No. NC3377701
Encompass_Preferred
All tubes have a frosted marking spot and engraved graduationsAttached flat top cap is easy to open and close with one handPart Numbers 230406 & 230407 feature a low retention surface
Catalog No. NC3377701 Supplier CELLTREAT Scientific Products Supplier No. 230407
All tubes have a frosted marking spot and engraved graduationsAttached flat top cap is easy to open and close with one handPart Numbers 230406 & 230407 feature a low retention surface