Learn More
Chemglass Life Sciences Tube, Micro Centrifuge, 1.5mL, Graduated, Polypropylene, SureSeal S, Assorted Colors, Packed in bags of 50 tubes, 10 bags per unit (2 bags of each color), Sterile
SDP
Catalog No. 501979481
Encompass_Preferred
Tube, Micro Centrifuge, 1.5mL, Graduated, Polypropylene, SureSeal S, Sterile, Assorted Colors, Packed in bags of 50 tubes, 10 bags per unit (2 bags of each color). SureSeal S microcentrifuge tubes. SureSeal S tubes are manufactured under clean room conditions, using only BioClear co-polymer, an advanced polypropylene that maximizes clarity, centrifugal strength (tested up to 28,000xg) and the usable temperature range (-90¿C to +121¿C). The oversized tube cap (with frosted marking surface), combined with the unique ring & groove closure, enhances the security of the liquid-tight seal, while minimizing the required locking forces for a more ergonomic opening/closing of the tube caps. All SureSeal S tubes are RNase-, DNase- and pyrogen-free.
Catalog No. 50-197-9481 Supplier Chemglass Life Sciences Supplier No. CLS4300C001
Tube, Micro Centrifuge, 1.5mL, Graduated, Polypropylene, SureSeal S, Sterile, Assorted Colors, Packed in bags of 50 tubes, 10 bags per unit (2 bags of each color). SureSeal S microcentrifuge tubes. SureSeal S tubes are manufactured under clean room conditions, using only BioClear co-polymer, an advanced polypropylene that maximizes clarity, centrifugal strength (tested up to 28,000xg) and the usable temperature range (-90¿C to +121¿C). The oversized tube cap (with frosted marking surface), combined with the unique ring & groove closure, enhances the security of the liquid-tight seal, while minimizing the required locking forces for a more ergonomic opening/closing of the tube caps. All SureSeal S tubes are RNase-, DNase- and pyrogen-free.