Learn More
Chemglass Life Sciences Micro Centrifuge Tube, 2.0mL, USP VI Grade Polypropylene, Graduated, Round Bottom, Sterile
SDP
Catalog No. 501897627
Encompass
• USP VI Grade PP material, free from heavy metal• RCF: 30,000xg• Endotoxin: 0.1EU• Clear graduations and frosted marking area• Special cap design for needle insertion• Easy opening and closing• Autoclave at 121 °C / 15 psi• Temperature range: -80°C to +121°C• DNase- and RNase-free, endotoxin levels of less than 0.1 EU• Packed in bags of 50
Catalog No. 50-189-7627 Supplier Chemglass Life Sciences Supplier No. CGE4302002
• USP VI Grade PP material, free from heavy metal• RCF: 30,000xg• Endotoxin: 0.1EU• Clear graduations and frosted marking area• Special cap design for needle insertion• Easy opening and closing• Autoclave at 121 °C / 15 psi• Temperature range: -80°C to +121°C• DNase- and RNase-free, endotoxin levels of less than 0.1 EU• Packed in bags of 50